Anti-Protein A Goat Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
200-1977
ROCK200-1977EA 71950 NOK
ROCK200-1977
Anti-Protein A Goat Polyclonal Antibody (Texas Red®)
Antistoffer
Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR