Anti-AKT2 Rat Monoclonal Antibody (Unconjugated) [clone: 16G11.A7]

Leverandør: Rockland Immunochemicals
200-301-096S
ROCK200-301-096SEA 2775 NOK
ROCK200-301-096S
Anti-AKT2 Rat Monoclonal Antibody (Unconjugated) [clone: 16G11.A7]
Antistoffer
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 16G11.A7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR