Anti-KLH Mouse Monoclonal Antibody (Unconjugated) [clone: 12B4.G3.A8]

Leverandør: Rockland Immunochemicals
200-301-466
ROCK200-301-466EA 6575 NOK
ROCK200-301-466
Anti-KLH Mouse Monoclonal Antibody (Unconjugated) [clone: 12B4.G3.A8]
Antistoffer
Anti-KLH can be used as a control in various assays or in ELISA assays to detect KLH or KLH conjugates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000

Type: Tag
Antigen: KLH
Clonality: Monoclonal
Clone: 12B4.G3.A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Klh,
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR