Anti-ATM Mouse Monoclonal Antibody (Unconjugated) [clone: 7C10D8]

Leverandør: Rockland Immunochemicals
200-301-500
ROCK200-301-500EA 11125 NOK
ROCK200-301-500
Anti-ATM Mouse Monoclonal Antibody (Unconjugated) [clone: 7C10D8]
Antistoffer
This Anti-ATM Protein Kinase pS1981 (MOUSE) Monoclonal Antibody clone has been optimized for IHC, but may also be used for western blotting, flow cytometry, immunoprecipitation, or immunofluorescence microscopy.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 1:100 - 1:500;

Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 7C10D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol ATM
Antigen-navn ATM serine/threonine kinase
Environmentally Preferable
Antigen synonymer AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 7C10D8
Reaktivitet Human,Mouse
Vert Mouse
Gen-ID 472
Isotype
Western blot Yes
ELISA Yes
Immunutfelling Yes
Modifikasjon phospho-Ser1981
Immunogen This antibody was produced from a synthetic peptide S-L-A-F-E-E-G-Sp-Q-S-T-T-I-S-S corresponding to aa 1974-1988 of human ATM.
Format Antibody fraction
Skjema Liquid (sterile filtered)
Rensing Protein A purified
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon none
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR