Anti-ATM Mouse Polyclonal Antibody (Biotin) [clone: 10H11.E12]

Leverandør: Rockland Immunochemicals
200-306-400
ROCK200-306-400EA 12000 NOK
ROCK200-306-400
Anti-ATM Mouse Polyclonal Antibody (Biotin) [clone: 10H11.E12]
Antistoffer
This antibody was tested by ELISA and western blotting against both the native and recombinant forms of the Protein. This reagent may also be suitable other biotin-streptavidin based assays.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: 10H11.E12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR