Anti-GFP Mouse Monoclonal Antibody (DyLight® 549) [clone: 9F9.F9]

Leverandør: Rockland Immunochemicals
200-342-215
ROCK200-342-215EA 9725 NOK
ROCK200-342-215
Anti-GFP Mouse Monoclonal Antibody (DyLight® 549) [clone: 9F9.F9]
Antistoffer
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: ELISA: 1:20000-1:40000; FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: DyLight® 549
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR