Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 549) [clone: 18F3.H11]

Leverandør: Rockland Immunochemicals
200-342-269
ROCK200-342-269EA 9925 NOK
ROCK200-342-269
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 549) [clone: 18F3.H11]
Antistoffer
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000-1:20000; FLISA: >1:20000; Western Blot: 1:10000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 18F3.H11
Conjugation: DyLight® 549
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR