Anti-BSA Mouse Monoclonal Antibody (Unconjugated) [clone: 5F9]

Leverandør: Rockland Immunochemicals
201-301-033
ROCK201-301-033EA 11125 NOK
ROCK201-301-033
Anti-BSA Mouse Monoclonal Antibody (Unconjugated) [clone: 5F9]
Antistoffer
Anti-ALB antibody is suitable for western blotting and ELISA.  The antibody may be suitable for other applications, although no supporting data is available.

Recommended Dilutions: ELISA: 1:120000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: BSA
Clonality: Monoclonal
Clone: 5F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR