Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Rockland Immunochemicals
212-4202
ROCK212-4202EA 7250 NOK
ROCK212-4202
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Rat
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR