Anti-IgG (H+L) Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
302-4102
ROCK302-4102EA 5700 NOK
ROCK302-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Cat
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Cat
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Kyssabsorpsjon none
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR