Anti-GST Goat Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
600-142-200
ROCK600-142-200EA 9500 NOK
ROCK600-142-200
Anti-GST Goat Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR