Anti-KLH Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
600-401-466
ROCK600-401-466EA 13475 NOK
ROCK600-401-466
Anti-KLH Rabbit Polyclonal Antibody
Antistoffer
Anti-KLH Antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR