Anti-TLK1 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
600-401-469
ROCK600-401-469EA 10075 NOK
ROCK600-401-469
Anti-TLK1 Rabbit Polyclonal Antibody
Antistoffer
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 86 kDa, 89 kDa or 64 kDa in size corresponding to isoforms of TLk-1.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: TLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Orangutan, Dog
Bestill nå

SPESIFIKASJONER

Antigensymbol TLK1
Antigen-navn Tousled-like Kinase 1
Antigen synonymer PKU-beta
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Orangutan,Chimpanzee,Human,Dog
Vert Rabbit
Gen-ID 9874
Western blot Yes
ELISA Yes
Immunutfelling Yes
Epitop internal
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 737-751 of human Tlk1 (Tousled-like kinase) (isoform 1) Protein.
Aminosyretall 737 to 751
Format Antibody fraction
Skjema Liquid (sterile filtered)
Rensing affinity-purified
Konsentrasjon 0,60 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon none
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR