Anti-KLH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Rockland Immunochemicals
600-403-466
ROCK600-403-466EA 9500 NOK
ROCK600-403-466
Anti-KLH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Anti-KLH Antibody Peroxidase Conjugated antibody has been assayed against 1.0 µg of KLH in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:400,000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR