Anti-ACK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Rockland Immunochemicals
600-403-939
ROCK600-403-939EA 11150 NOK
ROCK600-403-939
Anti-ACK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
This affinity-purified antibody is suitable for use in ELISA and western blotting assays.  The antibody reacts specifically with acetylated lysine residues.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000-1:5000;

Type: Primary
Antigen: ACK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mammalian, Avian,
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR