Anti-KLH Rabbit Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
600-406-466
ROCK600-406-466EA 9500 NOK
ROCK600-406-466
Anti-KLH Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Anti-KLH Antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Anti-KLH antibody biotin conjugate is suitable for use with streptavidin reporter systems.

Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:5000 - 1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR