Anti-GST Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
600-442-200
ROCK600-442-200EA 9500 NOK
ROCK600-442-200
Anti-GST Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol GST
Antigen-navn Glutathione-S-Transferase
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ImmunoFluorescens Yes
ChIP Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR