Anti-HA Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
600-442-384
ROCK600-442-384EA 9500 NOK
ROCK600-442-384
Anti-HA Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR