Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
603-142-002
ROCK603-142-002EA 2625 NOK
ROCK603-142-002
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)
Antistoffer
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Chicken
Vert Goat
Isotype
Western blot Yes
ImmunoFluorescens Yes
Immunogen Chicken IgG, whole molecule
Format Antibody fraction
Skjema Lyophilized
Rensing purified by immunoaffinity chromatography
Kyssabsorpsjon No
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagringstemp –20 °C
Frakttemp. Ambient
Uttestet applikasjon Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR