Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
603-442-002-05
ROCK603-442-002-05EA 0 NOK
ROCK603-442-002-05
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken,
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Rat
Vert Rabbit
Isotype
Western blot Yes
ImmunoFluorescens Yes
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR