Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
605-4915
ROCK605-4915EA 4925 NOK
ROCK605-4915
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)
Antistoffer
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Goat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG F(ab')2
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Texas Red®
Reaktivitet Goat
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Immunogen Goat IgG F(ab')2 fragment
Format Antibody fraction
Skjema Lyophilized
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon Human
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR