Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
605-742-002-05
ROCK605-742-002-05EA 8750 NOK
ROCK605-742-002-05
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR