Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Rockland Immunochemicals
609-105-123
ROCK609-105-123EA 6450 NOK
ROCK609-105-123
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Secondary antibody reagents are ideal for western blotting, immunochemistry, ELISA, Fluorescence Microscopy, Flow Cytometry as well as other antibody detection methods. An ELISA second antibody is conjugated to HRP or alkaline phosphatase.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR