Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Leverandør: Rockland Immunochemicals
609-106-123
ROCK609-106-123EA 5350 NOK
ROCK609-106-123
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Antistoffer
Biotin Secondary antibody reagents are ideal for western blotting, immunochemistry and ELISA as well as other antibody detection methods.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Human
Vert Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Immunogen Human IgG whole molecule
Format Antibody fraction
Skjema Lyophilized
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR