Anti-IgG2a Goat Polyclonal Antibody (ATTO 550)

Leverandør: Rockland Immunochemicals
610-154-041
ROCK610-154-041EA 7575 NOK
ROCK610-154-041
Anti-IgG2a Goat Polyclonal Antibody (ATTO 550)
Antistoffer
Anti-Mouse IgG2a secondary antibody is suitable for ELISA, immunochemistry western blotting as well as other anti IgG2a antibody based assays.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: >1:10000; Immunochemistry: >1:5000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: ATTO 550
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG2a
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering ATTO 550
Reaktivitet Mouse
Vert Goat
Isotype
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
ChIP Yes
Uttestet applikasjon Fluoresence ELISA, Fluorescence Resonance Energy Transfer, Stimulated Emission Depletion Microscopy

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR