Anti-IgG3 Rabbit Polyclonal Antibody (Cy2®)

Leverandør: Rockland Immunochemicals
610-411-043
ROCK610-411-043EA 5750 NOK
ROCK610-411-043
Anti-IgG3 Rabbit Polyclonal Antibody (Cy2®)
Antistoffer
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:200-1:400; Flow Cytometry 1:200-1:400
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG3 γ
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR