Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
610-442-002-05
ROCK610-442-002-05EA 8475 NOK
ROCK610-442-002-05
Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Secondary
Antigen: IgG(Mouse)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Mouse
Vert Rabbit
Isotype
Western blot Yes
ImmunoFluorescens Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR