Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
610-742-002
ROCK610-742-002EA 2625 NOK
ROCK610-742-002
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 549)
Antistoffer
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR