Anti-IgG F(c) Donkey Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
611-709-003
ROCK611-709-003EA 4200 NOK
ROCK611-709-003
Anti-IgG F(c) Donkey Polyclonal Antibody (Texas Red®)
Antistoffer
Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(c)
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR