Anti-IgG F(c) Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
612-1103
ROCK612-1103EA 3350 NOK
ROCK612-1103
Anti-IgG F(c) Goat Polyclonal Antibody
Antistoffer
Secondary Goat Anti-IgG F(c) Reacts with Rat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR