Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)

Leverandør: Rockland Immunochemicals
612-142-002-05
ROCK612-142-002-05EA 8475 NOK
ROCK612-142-002-05
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 549)
Antistoffer
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR