Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)

Leverandør: Rockland Immunochemicals
705-4913
ROCK705-4913EA 5350 NOK
ROCK705-4913
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Antistoffer
Secondary Rabbit Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR