Anti-IgG F(ab')2 Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
709-1118
ROCK709-1118EA 4200 NOK
ROCK709-1118
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Antistoffer
Antibody Anti-Human IgG F(ab')2 is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG F(ab')2
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Antistoff fragment F(ab')2
Immunogen Anti-Human IgG was produced by repeated immunization with human IgG F(ab')2 fragment in goat.
Format Antibody fraction
Skjema Liquid (sterile filtered)
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon Bovine, Horse, Mouse, Rat
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR