Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Leverandør: Rockland Immunochemicals
811-1302
ROCK811-1302EA 6200 NOK
ROCK811-1302
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Antistoffer
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Peroxidase
Reaktivitet Rabbit
Vert Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Gelskifte Yes
Flowcytometri Yes
ChIP Yes
Nøytralisering Yes
Immunogen Anti-Rabbit IgG whole molecule was produced by repeated immunization with Rabbit IgG whole molecule in goat.
Format Antibody fraction
Skjema Lyophilized
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon none
Lagringsbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR