Anti-IgG (H+L) Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Rockland Immunochemicals
811-7002
ROCK811-7002EA 3800 NOK
ROCK811-7002
Anti-IgG (H+L) Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR