ModDetect™ Phosphorothioate Panel
Leverandør: Rockland Immunochemicals
KNA-100
ROCKKNA-100EA 0 NOK
ROCKKNA-100
ModDetect™ Phosphorothioate Panel
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
The set includes five unlabelled ModDetect™ phosphorothioate detection reagents, curated by binding characteristics, and three reporter molecules labelled with peroxidase, 488 λ fluorophore and 649 λ fluorophore.
The different binding characteristics create opportunities to generate highly optimised immunoassays independent of oligonucleotide sequence, as opposed to spending significant efforts to optimise assays to underperforming reagents.
Designed for use in various immunoassays, such as ELISA, IF, and IHC.
Store three reporter molecule vials at 4 °C or −20 °C prior to restoration. For extended storage aliquot contents and freeze at −20 °C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
Expiration date is one year from date of receipt.
This set includes the following components: PS03, PS04, PS05, PS07, PS09, and three reporter molecules.