SSPE Buffer, 20X concentrate, pH 7,4
Leverandør: Rockland Immunochemicals
MB-044
ROCKMB-044EA 2975 NOK
ROCKMB-044
SSPE Buffer, 20X concentrate, pH 7,4
Buffere Buffere, elektroforese Buffere, hybridisering
20X SSPE buffer (3,0 M sodium chloride, 0,2 M sodium hydrogen phosphate, 0,02 M EDTA, pH 7,4) - DEPC treated.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
NB: This product is for research use only and is not intended for therapeutic or diagnostic applications.