DNA (Deoxyribonucleic acid) (from salmon testes)

Leverandør: Rockland Immunochemicals

MB-103-1000
ROCKMB-103-1000EA 3575 NOK
ROCKMB-103-1000
DNA (Deoxyribonucleic acid) (from salmon testes)
DNA (Deoxyribonucleic acid)

DNA from salmon is commonly used in the preparation of pre-hybridisation and hybridisation solutions. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA, and DNase- and RNase-free (DEPC-treated) distilled, deionised water.

MDL nummer: MFCD00130922
CAS-nummer: 9007-49-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR