Chicken blood, sterile
Leverandør: Rockland Immunochemicals
R302-0050
ROCKR302-0050EA 3525 NOK
ROCKR302-0050
Chicken blood, sterile
Blod
Chicken whole blood aseptically collected. These cell suspensions are ideal for washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.