Bovine/Calf Blood in Heparin Sodium
Leverandør: Rockland Immunochemicals
R500-0100
ROCKR500-0100EA 4200 NOK
ROCKR500-0100
Bovine/Calf Blood in Heparin Sodium
Blod
Bovine/calf blood in heparin sodium.
  • Aseptically collected and prepared to order
  • Blood draw in anticoagulant
Bovine/calf whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Bovine/calf red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration,More Product Information