Tubing clamps, Rotilabo®
Leverandør: Roth Carl
1162.1
ROTH1162.1EA 0 NOK
ROTH1162.1
Tubing clamps, Rotilabo®
Klemmer Slangeklemmer
Tubing clamps made of PBT, coloured.
  • Adjustable up to total closing of tube
  • Can be operated with one hand
  • Sterilisable at 160 °C