Gas cylinder holder, Sekuroka®
Leverandør: Roth Carl
1513.1
ROTH1513.1EA 0 NOK
ROTH1513.1
Gas cylinder holder, Sekuroka®
Veggkrok
This gas cylinder holder is suitable for safe attachment of gas bottles up to 350 mm Ø. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 60 mm thickness.
  • Bore hole for bolt
  • Variable attachment with black nylon strap