Tetrahydropyran

Leverandør: Roth Carl

3073.1
ROTH3073.1EA 0 NOK
ROTH3073.1
Tetrahydropyran
Tetrahydropyran
Formel: C₅H₁₀O
MW: 86.13 g/mol
Kokepunkt: 88
Smeltepunkt: –44.2 °C
Tetthet: 0.881 g/cm³ (20 °C)
MDL nummer: MFCD00006585
CAS-nummer: 142-68-7
EINECS: 205-552-8
FN: 3271
ADR: 3 II
Merck Indeks: 12,09356

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR