Fuksin (syre)

Leverandør: Roth Carl

Synonymer: CI 42685

4268.5
ROTH4268.5EA 0 NOK
ROTH4268.5
Fuksin (syre)
Fuksin (syre)
Formel: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585,55 g/mol
Smeltepunkt: > 130 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013286
CAS-nummer: 3244-88-0
EINECS: 221-816-5
Merck Indeks: 12,00109

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR