Protection masks, Sekuroka®
Leverandør: Roth Carl
8401.1
ROTH8401.1EA 0 NOK
ROTH8401.1
Protection masks, Sekuroka®
Masker Munnbind
Sekuroka® odour binding protection mask is made from active carbon tissue.
  • Free of fibre glass
  • Aluminum nose support
  • Earloops
Suitable for medical use.
Forpakning: Individually wrapped in foil.