Quartz wool
Leverandør: Roth Carl
9208.2
ROTH9208.2EA 8650 NOK
ROTH9208.2
Quartz wool
Bomull og glassull
This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.
  • Chemically pure
  • Fibre size 5 to 30 µm
Forpakning: Packed in plastic bottles.