HygroSoft Software
Leverandør: ROTRONIC INSTRUMENTS
HS-AWCODE
ROTRHS-AWCODEEA 25725 NOK
ROTRHS-AWCODE
HygroSoft Software
Programvare Laboratoriesoftware
The HygroSoft software is compatible with all Rotronic products with USB, UART or ethernet interfaces and runs on all WINDOWS 10 operating systems.
  • Live display of current measured values
  • Configuration of instruments
  • Probe calibration and adjustment
  • AwQuick function
  • Measurement within 4 to 5 minutes
  • Viewing of measured values is very easy and user-friendly
  • The data is presented in both tabular and graphical formats
The HygroSoft software can be used to adjust the settings of Rotronic instruments and probes. Depending on the instrument and probe, theMore Product Information