Beaker tongs, stainless steel/rubber
Leverandør: RSG
043.290
RSGA043.290EA 435 NOK
RSGA043.290
Beaker tongs, stainless steel/rubber
Tenger
Stainless steel 18/10. All-purpose beaker tongs with rubber protected tips. With second diameter for holding small flasks and beakers.