Newborn calf serum
Leverandør: Biowest
S0750-500 S0750-100 S075H-100 S075H-500 S075G-500
S0750-500EA 1250 NOK
S0750-500 S0750-100 S075H-100 S075H-500 S075G-500
Newborn calf serum
Serum
Newborn Calf Serum (NBCS) originates from 3 to 10 day old calves. It is a high quality, cost effective replacement for Foetal Bovine Serum (FBS) and contains a somewhat higher proportion of proteins and immunoglobulin, but fewer growth factors. With proper cell adaptation, it has been shown to perform very well in cell culture applications where routine cell growth and cell culture maintenance is the objective.