Safranine O 85 Dye, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: CI 50240

MQ200

S2255-100G S2255-25G
S2255-100GEA 2360 NOK
S2255-100G S2255-25G
Safranine O 85 Dye, Sigma-Aldrich®
Safranine O

Safranin O is a cationic dye, which displays less significant metachromasia. It is used to detect any variations in articular disease. Safranin O binds specifically to polyanions. It detects the amount of proteoglycan present in cartilages. Safranin O is used as a marker for glycosaminoglycan depletion in osteoarthritis. Safranin is used as a counter stain in gram staining.


Safranin O has been used in the histological analysis of femurs. It has also been used to detect the presence of glycosaminoglycans in the inner body of meniscus structures.

Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Tetthet: 1.447 g/cm³ (22 °C)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6
EINECS: 207-518-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f29ccd4018b9465adf1fc92b9b655f20b34c695&vid=636e827f22f1223f7e95b953168d1120fb5a44ab&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Nitrogen 13.6 - 16.8 %
Dye Content ≥ 85 %
Solubility (Colour) Dark Red to Very Dark Red
Solubility (Turbidity)
1 mg/mL, H2O
Clear
Wavelength 530 - 534 nm
E1%
50% EtOH and H2O
1250 - 1650
Recommended Retest Period 4 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR